Assay Details
Gene Symbol: LOC5572428 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846731.1 | XP_021702423.1 | 11 - 12 | 2800 | 130 | |
GenBank mRNA | AY432915.1 | - | 1 - 2 | 238 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
