Assay Details
Gene Symbol: LOC5569863 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658709.2 | XP_001658759.1 | 3 | 881 | 90 | |
GenBank mRNA | DQ440248.1 | - | 2 | 721 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
