Assay Details
Gene Symbol: LOC5566651 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001650964.2 | XP_001651014.1 | 5 | 1296 | 97 | |
XM_021850915.1 | XP_021706607.1 | 5 | 1310 | 97 | ||
GenBank mRNA | AY431242.1 | - | 2 | 324 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
