Assay Details
Gene Symbol: LOC5571414 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653617.2 | XP_001653667.1 | 2 | 537 | 67 | |
XM_001653619.2 | XP_001653669.1 | 2 | 537 | 67 | ||
XM_021851540.1 | XP_021707232.1 | 2 | 530 | 67 | ||
GenBank mRNA | AY431643.1 | - | 2 | 423 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
