Assay Details
Gene Symbol: LOC110679863 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021855835.1 | XP_021711527.1 | 1 | 143 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021855835.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
