Assay Details
Gene Symbol: LOC5576559 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850178.1 | XP_021705870.1 | 10 - 11 | 5488 | 75 | |
XM_021850186.1 | XP_021705878.1 | 10 - 11 | 4084 | 75 | ||
XM_021850196.1 | XP_021705888.1 | 10 - 11 | 5488 | 75 | ||
XM_021850206.1 | XP_021705898.1 | 10 - 11 | 5488 | 75 |
