Assay Details
Gene Symbol: LOC5576559 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850178.1 | XP_021705870.1 | 9 - 10 | 5353 | 86 | |
XM_021850186.1 | XP_021705878.1 | 9 - 10 | 3949 | 86 | ||
XM_021850189.1 | XP_021705881.1 | 9 - 10 | 5353 | 86 | ||
XM_021850196.1 | XP_021705888.1 | 9 - 10 | 5353 | 86 | ||
XM_021850206.1 | XP_021705898.1 | 9 - 10 | 5353 | 86 | ||
XM_021850211.1 | XP_021705903.1 | 9 - 10 | 5353 | 86 | ||
XM_021850218.1 | XP_021705910.1 | 9 - 10 | 5353 | 86 | ||
XM_021850224.1 | XP_021705916.1 | 9 - 10 | 5353 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
