Assay Details
Gene Symbol: LOC5579655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648079.2 | XP_001648129.1 | 3 - 4 | 337 | 75 | |
XM_021852485.1 | XP_021708177.1 | 3 - 4 | 239 | 75 | ||
XM_021852486.1 | XP_021708178.1 | 3 - 4 | 166 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
