Assay Details
Gene Symbol: LOC5579655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648079.2 | XP_001648129.1 | 5 - 6 | 606 | 62 | |
XM_021852485.1 | XP_021708177.1 | 5 - 6 | 508 | 62 | ||
XM_021852486.1 | XP_021708178.1 | 5 - 6 | 435 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
