Assay Details
Gene Symbol: LOC5564570 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648866.2 | XP_001648916.2 | 1 - 2 | 331 | 71 | |
XM_021852806.1 | XP_021708498.1 | 2 - 3 | 253 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
