Assay Details
Gene Symbol: LOC5564570 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648866.2 | XP_001648916.2 | 2 - 3 | 515 | 100 | |
XM_021852806.1 | XP_021708498.1 | 3 - 4 | 437 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
