Assay Details
Gene Symbol: LOC5578258 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001663732.2 | XP_001663782.1 | 5 - 6 | 2049 | 75 | |
XM_021841793.1 | XP_021697485.1 | 6 - 7 | 1906 | 75 | ||
XM_021841794.1 | XP_021697486.1 | 5 - 6 | 1857 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
