Assay Details
Gene Symbol: LOC5568422 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652199.2 | XP_001652249.2 | 1 - 2 | 65 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001652199.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
