Assay Details
Gene Symbol: LOC5576442 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656080.2 | XP_001656130.2 | 2 - 3 | 392 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5576442 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656080.2 | XP_001656130.2 | 2 - 3 | 392 | 88 |
Set Membership: |
![]() ![]() ![]() ![]() |