Assay Details
Gene Symbol: LOC5576440 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656082.2 | XP_001656132.1 | 4 - 5 | 665 | 88 | |
XM_001656084.2 | XP_001656134.1 | 4 - 5 | 652 | 88 | ||
XM_011495371.2 | XP_011493673.1 | 4 - 5 | 612 | 88 | ||
XM_021846050.1 | XP_021701742.1 | 4 - 5 | 617 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
