Assay Details
Gene Symbol: LOC5574645 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661512.2 | XP_001661562.2 | 1 - 2 | 350 | 106 | |
XM_021844217.1 | XP_021699909.1 | 2 - 3 | 403 | 106 | ||
XM_021844218.1 | XP_021699910.1 | 2 - 3 | 491 | 106 |

More Information
Set Membership: |
![]() ![]() |
