Assay Details
Gene Symbol: LOC5574645 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661512.2 | XP_001661562.2 | 3 - 4 | 566 | 98 | |
XM_021844217.1 | XP_021699909.1 | 4 - 5 | 619 | 98 | ||
XM_021844218.1 | XP_021699910.1 | 4 - 5 | 707 | 98 |
