Assay Details
Gene Symbol: LOC5566122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839662.1 | XP_021695354.1 | 14 - 15 | 2778 | 77 | |
XM_021839663.1 | XP_021695355.1 | 13 - 14 | 2637 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
