Assay Details
Gene Symbol: LOC5566122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839662.1 | XP_021695354.1 | 4 - 5 | 957 | 89 | |
XM_021839663.1 | XP_021695355.1 | 3 - 4 | 816 | 89 | ||
XM_021839665.1 | XP_021695357.1 | 4 - 5 | 957 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
