Assay Details
Gene Symbol: LOC5574010 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021856309.1 | XP_021712001.1 | 7 | 3021 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5574010 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021856309.1 | XP_021712001.1 | 7 | 3021 | 66 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |