Assay Details
Gene Symbol: LOC110680521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021856318.1 | XP_021712010.1 | 2 | 1917 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC110680521 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021856318.1 | XP_021712010.1 | 2 | 1917 | 129 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |