Assay Details
Gene Symbol: KMO_ANOGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_315983.4 | XP_315983.2 | 6 - 7 | 1519 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
