Assay Details
Gene Symbol: 1275382 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001687731.1 | XP_001687783.1 | 3 - 4 | 1185 | 90 | |
XM_314619.4 | XP_314619.3 | 3 - 4 | 865 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
