Assay Details
Gene Symbol: TPX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_320690.3 | XP_320690.3 | 3 - 4 | 513 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
