Assay Details
Gene Symbol: Tpx-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001178023.1 | NP_001171494.1 | 88 | |||
NM_001178024.1 | NP_001171495.1 | 88 | ||||
GenBank mRNA | AY329358.1 | - | 88 | |||
BI507212.1 | - | 88 | ||||
DB738631.1 | - | 88 | ||||
HP519531.1 | - | 88 | ||||
HP548122.1 | - | 88 | ||||
HP574245.1 | - | 88 |
