Assay Details
Gene Symbol: Gtpx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001178022.1 | NP_001171493.1 | 79 | |||
GenBank mRNA | AY329359.1 | - | 79 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001178022.1 transcripts.
