Assay Details
Gene Symbol: Gtpx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001178021.1 | NP_001171492.1 | 141 | |||
NM_001178022.1 | NP_001171493.1 | 141 | ||||
GenBank mRNA | AY329359.1 | - | 141 | |||
HP476796.1 | - | 141 | ||||
HP544782.1 | - | 141 |
