Assay Details
Gene Symbol: Pban | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110712.2 | NP_001104182.1 | 86 | |||
GenBank mRNA | DQ199622.1 | - | 86 | |||
DQ869054.1 | - | 86 | ||||
HP497479.1 | - | 86 | ||||
HP559628.1 | - | 86 |
