Assay Details
Gene Symbol: Uqcr11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001190814.1 | NP_001177743.1 | 113 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001190814.1 transcripts.
