Assay Details
Gene Symbol: LOC411586 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP510415.1 | - | 73 | |||
HP510416.1 | - | 73 | ||||
HP510417.1 | - | 73 | ||||
HP510418.1 | - | 73 | ||||
HP510419.1 | - | 73 | ||||
HP510420.1 | - | 73 | ||||
HP510421.1 | - | 73 | ||||
HP510422.1 | - | 73 | ||||
HP510423.1 | - | 73 | ||||
HP510424.1 | - | 73 | ||||
HP510425.1 | - | 73 | ||||
HP510426.1 | - | 73 | ||||
HP510427.1 | - | 73 | ||||
HP510428.1 | - | 73 | ||||
HP510429.1 | - | 73 | ||||
HP510430.1 | - | 73 | ||||
HP540556.1 | - | 73 | ||||
HP551746.1 | - | 73 |
