Assay Details
Gene Symbol: LOC725111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | GQ444137.1 | - | 146 | |||
GQ444138.1 | - | 146 | ||||
GQ444139.1 | - | 146 | ||||
GQ444140.1 | - | 146 | ||||
GQ444141.1 | - | 146 | ||||
HP499579.1 | - | 146 |
