Assay Details
Gene Symbol: LOC409036 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP469467.1 | - | 109 | |||
HP476459.1 | - | 109 | ||||
HP476460.1 | - | 109 | ||||
HP503011.1 | - | 109 | ||||
HP513785.1 | - | 109 | ||||
HP513786.1 | - | 109 |
