Assay Details
Gene Symbol: ACSF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206973.2 | NP_001193902.2 | 117 | |||
GenBank mRNA | HP478007.1 | - | 117 | |||
HP478008.1 | - | 117 | ||||
HP546549.1 | - | 117 | ||||
HP567360.1 | - | 117 |
