Assay Details
Gene Symbol: Ef-1a-f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011628.2 | NP_001011628.1 | 75 | |||
GenBank mRNA | HP478552.1 | - | 75 | |||
HP524564.1 | - | 75 | ||||
HP529239.1 | - | 75 |
