Assay Details
Gene Symbol: LOC552480 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP481680.1 | - | 92 | |||
HP522473.1 | - | 92 | ||||
HP575607.1 | - | 92 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC552480 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP481680.1 | - | 92 | |||
HP522473.1 | - | 92 | ||||
HP575607.1 | - | 92 |