Assay Details
Gene Symbol: LOC409835 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP523908.1 | - | 104 | |||
HP544581.1 | - | 104 | ||||
HP544582.1 | - | 104 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC409835 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP523908.1 | - | 104 | |||
HP544581.1 | - | 104 | ||||
HP544582.1 | - | 104 |