Assay Details
Gene Symbol: LOC552266 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP475960.1 | - | 78 | |||
HP475961.1 | - | 78 | ||||
HP475962.1 | - | 78 | ||||
HP475964.1 | - | 78 | ||||
HP496549.1 | - | 78 | ||||
HP496550.1 | - | 78 | ||||
HP520967.1 | - | 78 | ||||
HP534875.1 | - | 78 | ||||
HP546823.1 | - | 78 | ||||
HP568848.1 | - | 78 |
