Assay Details
Gene Symbol: LOC100577945 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP502669.1 | - | 100 | |||
HP561080.1 | - | 100 | ||||
HP561082.1 | - | 100 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100577945 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP502669.1 | - | 100 | |||
HP561080.1 | - | 100 | ||||
HP561082.1 | - | 100 |