Assay Details
Gene Symbol: tra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265585.1 | NP_001252514.1 | 118 | |||
GenBank mRNA | HP512643.1 | - | 118 | |||
JQ518314.1 | - | 118 | ||||
JQ518315.1 | - | 118 | ||||
JQ518316.1 | - | 118 |
