Assay Details
Gene Symbol: LOC410970 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP524933.1 | - | 174 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC410970 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP524933.1 | - | 174 |