Assay Details
Gene Symbol: Jhe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011563.1 | NP_001011563.1 | 107 | |||
GenBank mRNA | AB083009.1 | - | 107 | |||
AY526235.1 | - | 107 | ||||
HP465711.1 | - | 107 |
