Assay Details
Gene Symbol: Gycbeta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011632.2 | NP_001011632.1 | 57 | |||
GenBank mRNA | AB181489.1 | - | 57 | |||
AY769960.1 | - | 57 | ||||
HP493417.1 | - | 57 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001011632.2 transcripts.
