Assay Details
Gene Symbol: Tbh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077824.1 | NP_001071292.1 | 121 | |||
GenBank mRNA | AB897810.1 | - | 175 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tbh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077824.1 | NP_001071292.1 | 121 | |||
GenBank mRNA | AB897810.1 | - | 175 |