Assay Details
Gene Symbol: Obp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011590.1 | NP_001011590.1 | 86 | |||
GenBank mRNA | AF166496.1 | - | 86 | |||
AF393494.1 | - | 86 | ||||
HP535697.1 | - | 86 |
