Assay Details
Gene Symbol: RpL32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011587.1 | NP_001011587.1 | 107 | |||
GenBank mRNA | AF441189.1 | - | 107 | |||
HP465684.1 | - | 107 | ||||
HP490625.1 | - | 107 |
