Assay Details
Gene Symbol: Tpx-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001170973.1 | NP_001164444.1 | 95 | |||
GenBank mRNA | BI510108.1 | - | 95 | |||
DB738012.1 | - | 95 | ||||
HP462574.1 | - | 95 | ||||
HP482267.1 | - | 95 | ||||
HP572937.1 | - | 95 |
