Assay Details
Gene Symbol: mRpL16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001185104.1 | NP_001172033.1 | 101 | |||
GenBank mRNA | BI514960.1 | - | 101 | |||
DB758384.1 | - | 101 | ||||
HP492393.1 | - | 101 |
