Assay Details
Gene Symbol: Cdc37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242514.1 | NP_001229443.1 | 100 | |||
GenBank mRNA | HP479169.1 | - | 100 | |||
HP496542.1 | - | 100 | ||||
HP524302.1 | - | 100 | ||||
HP571634.1 | - | 100 |
