Assay Details
Gene Symbol: Eif3h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242520.1 | NP_001229449.1 | 83 | |||
GenBank mRNA | HP481338.1 | - | 83 | |||
HP501225.1 | - | 83 | ||||
HP523525.1 | - | 83 | ||||
HP557377.1 | - | 83 |
