Assay Details
Gene Symbol: LPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_101377.5 | NP_172961.1 | 2 - 3 | 498 | 68 | |
GenBank mRNA | AB053950.1 | - | 1 - 2 | 216 | 68 | |
AB061407.1 | - | 2 - 3 | 419 | 68 | ||
AY087673.1 | - | 2 - 3 | 411 | 68 | ||
BT003912.1 | - | 2 - 3 | 411 | 68 | ||
BT004969.1 | - | 1 - 2 | 216 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
